Indan 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,3-Dihydro-1H-indene, Indan
B21442.AP
B21442.0F
B21442.AE
B21442.APEA
165
EUR
B21442.AP
B21442.0F
B21442.AE
Indan 95%
Indan
Purity: 95%
CAS No.: 496-11-7
Molecular Formula: C9H10
Formula Weight: 118.18
Flash Point: 50°(122°F)
Refractive Index: 1.5370
Melting Point: -51°
Boiling Point: 175-177°
Beilstein Reference: 1904376
Merck Reference: 14,4931
UN No.: 3295
MDL No.: MFCD00003795
Formel:
C₉H₁₀ Molecular Weight: 118,18 g/mol Siedepunkt: 175…177 °C Schmelzpunkt: –51 °C Dichte: 0,962 Flammpunkt: 50 °C (122 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003795 CAS-Nummer: 496-11-7 EINECS: 207-814-7 UN: 3295 ADR: 3,III Merck Index: 14,4931 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...