3-Isochinolinol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Hydroxyisoquinoline

L19428.03 L19428.06
L19428.03EA 69.4 EUR
L19428.03 L19428.06
3-Isochinolinol ≥99%
3-Isochinolinol

Purity: 99% CAS No.: 7651-81-2 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: ca 250° dec. Beilstein Reference: 113230

Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Schmelzpunkt: ∼ 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075524
CAS-Nummer: 7651-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR