6-Isochinolinol

Lieferant: CHEMPUR

Synonyme: 6-Hydroxyisoquinoline, Isoquinolin-6-ol

50-0052.1 50-0052.5
CHMP50-0052.1EA 0 EUR
CHMP50-0052.1 CHMP50-0052.5
6-Isochinolinol
6-Isochinolinol
Formel: C₉H₇NO
MDL: MFCD04114860
CAS-Nummer: 7651-82-3
EINECS: 231-612-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR