Praseodymtribromid, dry_ultra 99,99 REO basis

Lieferant: Thermo Fisher Scientific
Achtung


ampuled under argon

35714.04 35714.09
35714.04EA 215 EUR
35714.04 35714.09
Praseodymtribromid, dry_ultra 99,99 REO basis
Praseodymtribromid

Purity: 99.99% CAS No.: 13536-53-3 Molecular Formula: PrBr3 Formula Weight: 380.63 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049762

Formel: Br₃Pr
Molecular Weight: 380.62 g/mol
Schmelzpunkt: 691 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049762
CAS-Nummer: 13536-53-3
EINECS: 236-893-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR