Praseodymtrichlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
39386.06
39386.14
39386.06EA
150
EUR
39386.06
39386.14
Praseodymtrichlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymtrichlorid
Purity: 99.99%
CAS No.: 10361-79-2
Molecular Formula: PrCl3
Formula Weight: 247.27
Physical Form: Powder, Ampouled under argon
Density: 4.02
Melting Point: 786°
Boiling Point: 1700°
Merck Reference: 14,7712
MDL No.: MFCD00011176
Formel:
Cl₃Pr Molecular Weight: 247.27 g/mol Siedepunkt: 1700 °C Schmelzpunkt: 786 Dichte: 4.02 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011176 CAS-Nummer: 10361-79-2 EINECS: 233-794-4 Merck Index: 14,7712 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...