Praseodymtrifluorid
Lieferant: FLUOROCHEM
007228-10G
007228-50G
007228-100G
FLUO007228-10GEA
0
EUR
FLUO007228-10G
FLUO007228-50G
FLUO007228-100G
Praseodymtrifluorid
Praseodymtrifluorid
Formel:
F₃Pr Molecular Weight: 197,9 g/mol Schmelzpunkt: 1395 °C Dichte: 6,3 g/cm³ (20 °C) |
MDL:
MFCD00016287 CAS-Nummer: 13709-46-1 EINECS: 237-254-9 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...