Praseodymtrinitrat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Praseodym(III)nitrat

12909.09 12909.18 12909.30 12909.A1
12909.09EA 36.5 EUR
12909.09 12909.18 12909.30 12909.A1
Praseodymtrinitrat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymtrinitrat Hydrat

Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide


Purity: 99.9% CAS No.: 15878-77-0 Molecular Formula: Pr(NO3)3.xH2O Formula Weight: 326.92(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Pale green Odour: Odourless UN No.: 1477 MDL No.: MFCD00011175

Formel: N₃O₉Pr
Molecular Weight: 326,92 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011175
CAS-Nummer: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR