Lanthan(III)oxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

41644.18 41644.30
41644.18EA 245 EUR
41644.18 41644.30
Lanthan(III)oxid ≥99,9% (Metall-Basis)
Lanthan(III)oxid

Purity: 99.9% CAS No.: 1312-81-8 Molecular Formula: La2O3 Formula Weight: 325.82 Physical Form: 3-12mm Sintered Lump Density: 6.51 Melting Point: 2307° Boiling Point: 4200° Solubility: Insoluble in water. Soluble in dilute mineral acids Merck Reference: 14,5363 MDL No.: MFCD00011071


For vacuum deposition

Formel: La₂O₃
Molecular Weight: 325.81 g/mol
Siedepunkt: 4200 °C
Schmelzpunkt: 2307 °C
Dichte: 6.51
Lagertemperatur: Raumtemperatur
MDL: MFCD00011071
CAS-Nummer: 1312-81-8
EINECS: 215-200-5
Merck Index: 14,5363

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR