Praseodym(III,IV)oxid 99,996 REO basis, REacton®
Lieferant: Thermo Fisher Scientific
Achtung
10914.06 10914.14
10914.06EA 71.9 EUR
10914.06 10914.14
Praseodym(III,IV)oxid 99,996 REO basis, REacton®
Praseodym(III,IV)oxid
Purity: 99.996% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178
Formel: O₁₁Pr₆
Molecular Weight: 1021.44 g/mol
Schmelzpunkt: 2500 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011178
CAS-Nummer: 12037-29-5
EINECS: 234-857-9