Praseodym(III,IV)oxid 99,5
Lieferant: Thermo Fisher Scientific
|
Achtung
|
11234.14
11234.22
11234.A1
11234.14EA
103
EUR
11234.14
11234.22
11234.A1
Praseodym(III,IV)oxid 99,5
Praseodym(III,IV)oxid
Purity: 99.5%
CAS No.: 12037-29-5
Molecular Formula: Pr6O11
Formula Weight: 1021.43
Physical Form: Powder
Melting Point: 2500°
MDL No.: MFCD00011178
Formel:
O₁₁Pr₆ Molecular Weight: 1021.44 g/mol Schmelzpunkt: 2500 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011178 CAS-Nummer: 12037-29-5 EINECS: 234-857-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...