Praseodym(III,IV)oxid 99 REO basis

Lieferant: Thermo Fisher Scientific
Achtung

44594.30 44594.A1
44594.30EA 131 EUR
44594.30 44594.A1
Praseodym(III,IV)oxid 99 REO basis
Praseodym(III,IV)oxid

Purity: 99% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178

Formel: O₁₁Pr₆
Molecular Weight: 1021.44 g/mol
Schmelzpunkt: 2500 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011178
CAS-Nummer: 12037-29-5
EINECS: 234-857-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR