Silberbromid ≥99,5%
Lieferant: Thermo Fisher Scientific
Purity: 99.9%
CAS No.: 7785-23-1
Molecular Formula: AgBr
Formula Weight: 187.78
Physical Form: Powder
Density: 6.47
Appearance: Pale yellow
Odour: Odourless
Melting Point: 432°
Solubility: Practically insoluble in water. Insoluble in alcohol and acids. Partially soluble in ammonia
Merck Reference: 14,8506
UN No.: 3077
MDL No.: MFCD00003398
Formel:
AgBr Siedepunkt: 1300 °C (1013 hPa) Schmelzpunkt: 432 °C Dichte: 6,47 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003398 CAS-Nummer: 7785-23-1 EINECS: 232-076-8 ADR: 9,III Merck Index: 14,8506 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...