Silberfluorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Silbermonofluorid

11609.06 11609.14
11609.06EA 109 EUR
11609.06 11609.14
Silberfluorid ≥98%
Silberfluorid

Purity: 98% CAS No.: 7775-41-9 Molecular Formula: AgF Formula Weight: 126.87 Physical Form: Crystalline Density: 5.852 Appearance: Orange/brown Melting Point: 435° Boiling Point: ca 1150° Solubility: Soluble in HF, ammonia, CH3CN Merck Reference: 14,8514 UN No.: 3260 MDL No.: MFCD00003410

Formel: AgF
Molecular Weight: 126.87 g/mol
Siedepunkt: ∼1150 °C
Schmelzpunkt: 435 °C
Dichte: 5.852
Lagertemperatur: Raumtemperatur
MDL: MFCD00003410
CAS-Nummer: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8 II
Merck Index: 12,08657

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR