Kupferselenid ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Copper(II) selenide
17901.06
17901.14
17901.06EA
70.1
EUR
17901.06
17901.14
Kupferselenid ≥99,5% (Metall-Basis)
Kupferselenid
Purity: 99.5%
CAS No.: 1317-41-5
Molecular Formula: CuSe
Formula Weight: 142.50
Physical Form: 6mm & down pieces
Density: 6.0-6.6
Melting Point: 550° dec.
Solubility: Soluble in HCl with H2Se evolution, in H2SO4 with SO2 evolution. Oxidized to CuSeO3 by HNO3
Merck Reference: 14,2651
UN No.: 3077
MDL No.: MFCD00016058
In semiconductors, as catalyst in Kjeldahl digestion
Formel:
CuSe Molecular Weight: 142,51 g/mol Schmelzpunkt: 550 °C (Zers.) Dichte: 6,0…6.6 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016058 CAS-Nummer: 1317-41-5 EINECS: 215-272-8 UN: 3077 ADR: 9,III Merck Index: 14,2651 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...