Zinkoxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Zink(II)oxid

44264.22 44264.14
44264.22EA 320 EUR
44264.22 44264.14
Zinkoxid ≥99,9% (Metall-Basis)
Zinkoxid

Purity: 99.9% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 10-12mm dia x 4-5mm thick, Tablets, ≈2.3g/tablet Odour: Odourless Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Formel: ZnO
Molecular Weight: 81,39 g/mol
Schmelzpunkt: 1975 °C
Dichte: 5,61 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011300
CAS-Nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR