Zinkoxid ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Zink(II)oxid
87812.18
87812.30
87812.18EA
33.7
EUR
87812.18
87812.30
Zinkoxid ≥99,99% (Metall-Basis)
Zinkoxid
Purity: 99.99%
CAS No.: 1314-13-2
Molecular Formula: ZnO
Formula Weight: 81.37
Physical Form: Powder
Density: 5.606
Melting Point: 1975°
Merck Reference: 14,10147
UN No.: 3077
MDL No.: MFCD00011300
Formel:
ZnO Schmelzpunkt: 1975 °C Dichte: 5,606 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011300 CAS-Nummer: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 14,10147 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...