Zinkperoxid (∼50% ZnO₂)

Lieferant: Thermo Fisher Scientific
Gefahr

39635.22 39635.36
39635.22EA 36.7 EUR
39635.22 39635.36
Zinkperoxid (∼50% ZnO₂)
Zinkperoxid

CAS No.: 1314-22-3 Molecular Formula: ZnO2 Formula Weight: 97.38 Physical Form: Powder, remainder ZnO Odour: Odourless Melting Point: 150° dec. UN No.: 1516 MDL No.: MFCD00137469


Curing agent, rubber accelerator

Formel: O₂Zn
Molecular Weight: 97.39 g/mol
Schmelzpunkt: >150 °C (Zers.)
Dichte: 1.57 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00137469
CAS-Nummer: 1314-22-3
EINECS: 215-226-7
UN: 1516
ADR: 5.1 II
Merck Index: 12,10282

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR