Quecksilberselenid ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
12832.03
12832.06
12832.03EA
81.2
EUR
12832.03
12832.06
Quecksilberselenid ≥99,999% (Metall-Basis)
Quecksilberselenid
Purity: 99.999%
CAS No.: 20601-83-6
Molecular Formula: HgSe
Formula Weight: 279.55
Physical Form: -100 Mesh Powder
Density: 8.21
Appearance: Gray
Odour: Odourless
Melting Point: subl.
UN No.: 2025
MDL No.: MFCD00049743
Formel:
HgSe Molecular Weight: 279,55 g/mol Schmelzpunkt: (Subl.) Dichte: 8,21 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049743 CAS-Nummer: 20601-83-6 EINECS: 243-910-5 UN: 2025 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...