Zinkselenid ≥99,995% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Zinc (II) selenide

13241.18 13241.30
13241.18EA 123 EUR
13241.18 13241.30
Zinkselenid ≥99,995% (Metall-Basis)
Zinkselenid

Purity: 99.995% CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Physical Form: 3-12mm Lump Density: 5.42 Melting Point: 1100° Merck Reference: 14,10156 UN No.: 3283 MDL No.: MFCD00011296

Formel: SeZn
Molecular Weight: 144,35 g/mol
Schmelzpunkt: >1100 °C
Dichte: 5,42
Lagertemperatur: Raumtemperatur
MDL: MFCD00011296
CAS-Nummer: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 14,10156

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR