Cadmium(II)tellurid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cadmium telluride
14367.06
14367.14
14367.06EA
61.3
EUR
14367.06
14367.14
Cadmium(II)tellurid ≥99,999% (Metall-Basis)
Cadmium(II)tellurid
Purity: 99.999%
CAS No.: 1306-25-8
Molecular Formula: CdTe
Formula Weight: 240.00
Physical Form: -325 Mesh Polycrystalline Powder
Density: 6.20
Appearance: Black
Odour: Odourless
Melting Point: 1041°
Solubility: Insoluble in water and acids except HNO3 in which it is soluble with decomposition
Merck Reference: 14,1629
UN No.: 3077
MDL No.: MFCD00015998
Semiconductors, phosphors
Formel:
CdTe Molecular Weight: 240.01 g/mol Schmelzpunkt: 1041 °C Dichte: 6.20 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015998 CAS-Nummer: 1306-25-8 EINECS: 215-149-9 UN: 3077 ADR: 9 III Merck Index: 14,1629 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...