Zirkonium(IV)fluorid ≥99% hafniumarm
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Zirkontetrafluorid
11542.14
11542.22
11542.14EA
79.4
EUR
11542.14
11542.22
Zirkonium(IV)fluorid ≥99% hafniumarm
Zirkonium(IV)fluorid
Purity: 99%
CAS No.: 7783-64-4
Molecular Formula: F4Zr
Formula Weight: 167.21
Physical Form: Powder
Density: 4.43
Refractive Index: 1.59
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 600° subl.
Merck Reference: 14,10175
UN No.: 3260
MDL No.: MFCD00011307
Formel:
F₄Zr Molecular Weight: 167,22 g/mol Schmelzpunkt: 600 °C (Subl.) Dichte: 4,43 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011307 CAS-Nummer: 7783-64-4 EINECS: 232-018-1 UN: 3260 ADR: 8,II Merck Index: 14,10175 |