Zirkonium(IV)fluorid
Lieferant: FLUOROCHEM
Synonyme:
Zirkontetrafluorid
007787.25G
007787-100G
FLUO007787.25GEA
0
EUR
FLUO007787.25G
FLUO007787-100G
Zirkonium(IV)fluorid
Zirkonium(IV)fluorid
Formel:
F₄Zr Molecular Weight: 167,22 g/mol Dichte: 4,43 g/cm³ (20 °C) |
MDL:
MFCD00011307 CAS-Nummer: 7783-64-4 EINECS: 232-018-1 UN: 3260 ADR: 8,II Merck Index: 12,10308 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...