Hafnium(IV)tellurid ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Hafnium telluride
39223.03
39223.06
39223.03EA
121
EUR
39223.03
39223.06
Hafnium(IV)tellurid ≥99,5% (Metall-Basis)
Hafnium(IV)tellurid
Purity: 99.5%
CAS No.: 39082-23-0
Molecular Formula: HfTe2
Formula Weight: 433.69
Physical Form: -325 Mesh Powder
Formel:
HfTe₂ Molecular Weight: 433,69 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049947 CAS-Nummer: 39082-23-0 EINECS: 254-280-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...