Chrom(VI)oxid 98+% ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Chromtrioxid, Chromsäureanhydrid
36468.22
36468.36
36468.22EA
50.2
EUR
36468.22
36468.36
Chrom(VI)oxid 98+% ACS
Chrom(VI)oxid
Purity: 98+%
CAS No.: 1333-82-0
Molecular Formula: CrO3
Formula Weight: 99.99
Physical Form: Crystalline
Density: 2.7
Appearance: Red
Melting Point: 196°
Boiling Point: 250° dec.
Solubility: Soluble in water, H2SO4, and alcohol
Merck Reference: 14,2235
UN No.: 1463
MDL No.: MFCD00010952
Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, Coloured glass, oxidizing agent in organic chemistry
Formel:
CrO₃ Molecular Weight: 99,99 g/mol Siedepunkt: 250 °C (Zersetzung) Schmelzpunkt: 196 °C Dichte: 2,7 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010952 CAS-Nummer: 1333-82-0 EINECS: 215-607-8 UN: 1463 ADR: 5.1,II REACH: 01-2119458868-17 Merck Index: 14,2235 |