1-Bromnaphthalin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
α-Naphthylbromid, α-Bromnaphthalin
A14122.30
A14122.0B
A14122.18
A14122.30EA
63.1
EUR
A14122.30
A14122.0B
A14122.18
1-Bromnaphthalin ≥97%
1-Bromnaphthalin
Purity: 97%
CAS No.: 90-11-9
Molecular Formula: C10H7Br
Formula Weight: 207.08
Flash Point:
Formel:
C₁₀H₇Br Molecular Weight: 207,07 g/mol Siedepunkt: 279…281 °C Schmelzpunkt: –2…+0 °C Dichte: 1,485 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003868 CAS-Nummer: 90-11-9 EINECS: 201-965-2 Merck Index: 14,1425 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...