2-Bromnaphthalin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: β-Naphthylbromid

A15300.06 A15300.14 A15300.22
A15300.06EA 33.1 EUR
A15300.06 A15300.14 A15300.22
2-Bromnaphthalin ≥98%
2-Bromnaphthalin

Purity: 98+% CAS No.: 580-13-2 Molecular Formula: C10H7Br Formula Weight: 207.08 Flash Point: 112°(233°F) Odour: Odourless Boiling Point: 281-282° Beilstein Reference: 1858110 Merck Reference: 14,1426 MDL No.: MFCD00004051

Formel: C₁₀H₇Br
Molecular Weight: 207,07 g/mol
Siedepunkt: 281…282 °C
Schmelzpunkt: 53…57 °C
Dichte: 1,605
Flammpunkt: 112 °C (233 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004051
CAS-Nummer: 580-13-2
EINECS: 209-452-5
Merck Index: 14,1426

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR