1-Brom-4-fluornaphthalin

Lieferant: FLUOROCHEM
Achtung

CFF850. 004229-25G 004229-100G
CHMPCFF850.EA 0 EUR
CHMPCFF850. FLUO004229-25G FLUO004229-100G
1-Brom-4-fluornaphthalin
1-Bromo-4-fluoronaphthalene
Formel: C₁₀H₆BrF
MDL: MFCD00051473
CAS-Nummer: 341-41-3
EINECS: 206-434-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR