2,6-Naphthalindicarbonsäure

Lieferant: CHEMPUR

Synonyme: Naphthalin-2,6-dicarbonsäure

FL13171.POR 013171-100G 013171-25G 013171-5G
CHMPFL13171.POREA 0 EUR
CHMPFL13171.POR FLUO013171-100G FLUO013171-25G FLUO013171-5G
2,6-Naphthalindicarbonsäure
2,6-Naphthalindicarbonsäure
Formel: C₁₂H₈O₄
Molecular Weight: 216.19 g/mol
Siedepunkt: 422
Schmelzpunkt: 366…368 °C
MDL: MFCD00004105
CAS-Nummer: 1141-38-4
EINECS: 214-527-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR