1-Fluornaphthalin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A18378.06
A18378.14
A18378.22
A18378.06EA
40.8
EUR
A18378.06
A18378.14
A18378.22
1-Fluornaphthalin ≥98%
1-Fluoronaphthalene
Purity: 98%
CAS No.: 321-38-0
Molecular Formula: C10H7F
Formula Weight: 146.17
Flash Point: 65°(149°F)
Refractive Index: 1.5930
Melting Point: -10°
Boiling Point: 215-217°
Beilstein Reference: 1906413
MDL No.: MFCD00003873
Formel:
C₁₀H₇F Molecular Weight: 146,16 g/mol Siedepunkt: 215…217 °C Schmelzpunkt: –10 °C Dichte: 1,331 Flammpunkt: 65 °C (149 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003873 CAS-Nummer: 321-38-0 EINECS: 206-287-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...