Octafluornaphthalin 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Perfluoronaphthalene
B21699.03
B21699.06
B21699.14
B21699.03EA
52.4
EUR
B21699.03
B21699.06
B21699.14
Octafluornaphthalin 96%
Octafluornaphthalin
Purity: 96%
CAS No.: 313-72-4
Molecular Formula: C10F8
Formula Weight: 272.10
Melting Point: 84-89°
Boiling Point: 209°
MDL No.: MFCD00014307
Formel:
C₁₀F₈ Molecular Weight: 272.1 g/mol Siedepunkt: 209 °C Schmelzpunkt: 84…89 °C Dichte: 2.069 g/cm³ (–70 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014307 CAS-Nummer: 313-72-4 EINECS: 206-239-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...