Octafluornaphthalin
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
Perfluoronaphthalene
003270-1G
003270-5G
003270-25G
FLUO003270-1GEA
42.9
EUR
FLUO003270-1G
FLUO003270-5G
FLUO003270-25G
Octafluornaphthalin
Octafluornaphthalin
Formel:
C₁₀F₈ Molecular Weight: 272.1 g/mol Siedepunkt: 238 Schmelzpunkt: 80…86 °C Dichte: 2.069 g/cm³ (–70 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014307 CAS-Nummer: 313-72-4 EINECS: 206-239-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...