2-Naphthol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxynaphthalin, β-Naphthol, β-Hydroxynaphthalin

A14564.0B A14564.0I A14564.30
A14564.0BEA 76.5 EUR
A14564.0B A14564.0I A14564.30
2-Naphthol ≥98%
2-Naphthol

Purity: 98+% CAS No.: 135-19-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 153°(307°F) Odour: Phenol-like pH: 5.77 Melting Point: 120-124° Boiling Point: 285-286° Beilstein Reference: 742134 Merck Reference: 14,6384 UN No.: 3077 MDL No.: MFCD00004067

Formel: C₁₀H₈O
Molecular Weight: 144.17 g/mol
Siedepunkt: 285…286 °C
Schmelzpunkt: 120…124 °C
Dichte: 1.280
Flammpunkt: 153 °C (307 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004067
CAS-Nummer: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9 III
Merck Index: 12,06471

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR