1-(Chlormethyl)naphthalin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
α-Naphtylmethylchlorid
109450050
109451000
109455000
ACRO109450050EA
28.9
EUR
ACRO109450050
ACRO109451000
ACRO109455000
1-(Chlormethyl)naphthalin 95%
1-(Chlormethyl)naphthalin
Formel:
C₁₁H₉Cl Molecular Weight: 176.65 g/mol Siedepunkt: 291 Schmelzpunkt: 31…31 °C Dichte: 1.17 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004042 CAS-Nummer: 86-52-2 EINECS: 201-678-2 UN: 3261 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | Clear yellow to brown Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.6350 to 1.6390 (20°C, 589 nm) |
HPLC | Remainder 2-chloromethylnaphtalene |
Acid value | ≤35 mg KOH/g |
Peroxide value | ≤4 meq O2/kg |
UV | at 280 nm A: ≤0.1 |
Residue after ign. (sulfated) | ≤0.3 % |
Specific optical rotation | (after 24 h) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...