2-Methyl-1-nitronaphthalin

Lieferant: CHEMPUR

XBX00241.250 XBX00241.1
CHMPXBX00241.250EA 0 EUR
CHMPXBX00241.250 MAYBXBX00241.1
2-Methyl-1-nitronaphthalin
2-Methyl-1-nitronaphthalene
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2 g/mol
Siedepunkt: 318 °C (1013 hPa)
Schmelzpunkt: 81 °C
MDL: MFCD00003914
CAS-Nummer: 881-03-8
EINECS: 212-917-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR