1-Methylnaphthalin 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: α-Methylnaphthalin

A15327.14 A15327.22 A15327.36
A15327.14EA 25.5 EUR
A15327.14 A15327.22 A15327.36
1-Methylnaphthalin 96%
1-Methylnaphthalin

Purity: 96% CAS No.: 90-12-0 Molecular Formula: C11H10 Formula Weight: 142.20 Flash Point: 82°(179°F) Refractive Index: 1.6150 Melting Point: -22° Boiling Point: 240-243° Beilstein Reference: 506793 UN No.: 3082 MDL No.: MFCD00004034

Formel: C₁₁H₁₀
Molecular Weight: 142,2 g/mol
Siedepunkt: 240…243 °C
Schmelzpunkt: –22 °C
Dichte: 1,024
Flammpunkt: 82 °C (179 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004034
CAS-Nummer: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR