3,5-Dibrompyridin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A10875.06
A10875.14
A10875.22
A10875.06EA
67
EUR
A10875.06
A10875.14
A10875.22
3,5-Dibrompyridin ≥98%
3,5-Dibromopyridine
Purity: 98+%
CAS No.: 625-92-3
Molecular Formula: C5H3Br2N
Formula Weight: 236.90
Melting Point: 110-113°
Beilstein Reference: 108477
Formel:
C₅H₃Br₂N Molecular Weight: 236,89 g/mol Siedepunkt: 222 °C (1013 hPa) Schmelzpunkt: 110…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014634 CAS-Nummer: 625-92-3 EINECS: 210-916-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...