3-Brom-2-chlorpyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
AC36065.1
AC36065.10
360650050
360650250
MAYBAC36065.1EA
0
EUR
MAYBAC36065.1
MAYBAC36065.10
ACRO360650050
ACRO360650250
3-Brom-2-chlorpyridin 98%
3-Bromo-2-chloropyridine
Formel:
C₅H₃BrClN Molecular Weight: 192.44 g/mol Siedepunkt: 97 °C (1.3kPa) Schmelzpunkt: 57.9…58.3 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234007 CAS-Nummer: 52200-48-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or crystals or flakes or solid |
Infrared spectrum | Conforms |
Melting point | 53 to 59 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...