2-Brom-3-nitropyridin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L20019.03 L20019.06
L20019.03EA 27.2 EUR
L20019.03 L20019.06
2-Brom-3-nitropyridin ≥98%
2-Bromo-3-nitropyridine

Purity: 98% CAS No.: 19755-53-4 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Melting Point: 122-125° Beilstein Reference: 124504

Formel: C₅H₃BrN₂O₂
Molecular Weight: 203 g/mol
Schmelzpunkt: 122…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00955613
CAS-Nummer: 19755-53-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR