2,3,4,6-Tetrachlorpyridin

Lieferant: Thermo Scientific

BTB15033.250 BTB15033.1
MAYBBTB15033.250EA 0 EUR
MAYBBTB15033.250 MAYBBTB15033.1
2,3,4,6-Tetrachlorpyridin
2,3,4,6-Tetrachloropyridine
Formel: C₅HCl₄N
Molecular Weight: 216,88 g/mol
MDL: MFCD01312169
CAS-Nummer: 14121-36-9
EINECS: 237-973-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR