2,4-Dichlorpyridin
Lieferant: FLUOROCHEM
011684-5G
011684-25G
FLUO011684-5GEA
36.9
EUR
FLUO011684-5G
FLUO011684-25G
2,4-Dichlorpyridin
2,4-Dichloropyridine
Formel:
C₅H₃Cl₂N Molecular Weight: 147,99 g/mol |
MDL:
MFCD00955618 CAS-Nummer: 26452-80-2 EINECS: 247-717-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...