2,6-Dichlorpyridin ≥98%

Lieferant: Thermo Scientific
Gefahr

A11364.0E A11364.22 A11364.36
A11364.0EEA 552 EUR
A11364.0E A11364.22 A11364.36
2,6-Dichlorpyridin ≥98%
2,6-Dichloropyridine

Purity: 98% CAS No.: 2402-78-0 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Odour: Unpleasant Melting Point: 84-88° Boiling Point: 211° Beilstein Reference: 108664 UN No.: 2811 MDL No.: MFCD00006244

Formel: C₅H₃Cl₂N
Molecular Weight: 147,99 g/mol
Siedepunkt: 211 °C
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006244
CAS-Nummer: 2402-78-0
EINECS: 219-282-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR