3,5-Dichlorpyridin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L03380.14
L03380.22
L03380.14EA
38
EUR
L03380.14
L03380.22
3,5-Dichlorpyridin ≥98%
3,5-Dichloropyridine
Purity: 98%
CAS No.: 2457-47-8
Molecular Formula: C5H3Cl2N
Formula Weight: 147.99
Flash Point: 74°(165°F)
Odour: Unpleasant
Boiling Point: 178°
Beilstein Reference: 1973
UN No.: 2811
MDL No.: MFCD00006376
Formel:
C₅H₃Cl₂N Molecular Weight: 147,99 g/mol Siedepunkt: 178 °C Schmelzpunkt: 63…67 °C Dichte: 1,39 Flammpunkt: 74 °C (165 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006376 CAS-Nummer: 2457-47-8 EINECS: 219-537-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...