2-Chlor-5-nitropyridin
Lieferant: FLUOROCHEM
|
Achtung
|
010698-5G
010698-25G
010698-100G
FLUO010698-5GEA
0
EUR
FLUO010698-5G
FLUO010698-25G
FLUO010698-100G
2-Chlor-5-nitropyridin
2-Chloro-5-nitropyridine
Formel:
C₅H₃ClN₂O₂ Schmelzpunkt: 107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006240 CAS-Nummer: 4548-45-2 EINECS: 224-908-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...