3-Aminopyridin 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Pyridylamin, Pyridin-3-amin
104560250
104561000
104565000
ACRO104560250EA
28.9
EUR
ACRO104560250
ACRO104561000
ACRO104565000
3-Aminopyridin 99%
3-Aminopyridin
Formel:
C₅H₆N₂ Molecular Weight: 94,12 g/mol Siedepunkt: ∼248 °C (1013 hPa) Schmelzpunkt: 60…63 °C Flammpunkt: 124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006400 CAS-Nummer: 462-08-8 EINECS: 207-322-2 UN: 2671 ADR: 6.1,II Merck Index: 13,00472 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Crystals or flakes or crystalline mass |
Infrared spectrum | Conforms |
Melting point | 60 to 63 °C |
GC | ≥98.5 % |
Moisture | ≤3.0 % (as packaged) |
Viscosity | 0.2 to 0.6 s |
Free acid (CH3COOH) | ≤0.06 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...