3-Aminopyridin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Pyridylamin, Pyridin-3-amin

A13216.0B A13216.18 A13216.30
A13216.0BEA 410 EUR
A13216.0B A13216.18 A13216.30
3-Aminopyridin 99%
3-Aminopyridin

Purity: 99% CAS No.: 462-08-8 Molecular Formula: C5H6N2 Formula Weight: 94.12 Flash Point: 88°(190°F) Odour: Unpleasant pH: 10 Melting Point: 58-63° Boiling Point: 250-252° Beilstein Reference: 105692 Merck Reference: 14,473 UN No.: 2671 MDL No.: MFCD00006400

Formel: C₅H₆N₂
Molecular Weight: 94,12 g/mol
Siedepunkt: 250…252 °C
Schmelzpunkt: 58…63 °C
Flammpunkt: 88 °C (190 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006400
CAS-Nummer: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Merck Index: 14,473

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR