3,4-Diaminopyridin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,4-DAP, 3,4-Pyridindiamin, Pyridin-3,4-diyldiamin

A12465.03 A12465.06 A12465.14
A12465.03EA 35.1 EUR
A12465.03 A12465.06 A12465.14
3,4-Diaminopyridin ≥98%
3,4-Diaminopyridin

Purity: 98+% CAS No.: 54-96-6 Molecular Formula: C5H7N3 Formula Weight: 109.13 Flash Point: 240°(464°F) Odour: Unpleasant Beilstein Reference: 110232 Merck Reference: 14,2986 UN No.: 2811 MDL No.: MFCD00006401

Formel: C₅H₇N₃
Molecular Weight: 109,13 g/mol
Siedepunkt: 262 °C (1013 hPa)
Schmelzpunkt: 217…220 °C
Flammpunkt: 240 °C (464 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006401
CAS-Nummer: 54-96-6
EINECS: 200-220-9
UN: 2811
ADR: 6.1,II
Merck Index: 14,2986

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR