2-Brom-3-methylpyridin 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Bromo-3-picoline
AC35766.1
AC35766.10
AC35766.50
357660050
MAYBAC35766.1EA
0
EUR
MAYBAC35766.1
MAYBAC35766.10
MAYBAC35766.50
ACRO357660050
2-Brom-3-methylpyridin 95%
2-Brom-3-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172,02 g/mol Flammpunkt: >110 °C |
MDL:
MFCD00239380 CAS-Nummer: 3430-17-9 |
Spezifikation Testergebnisse
Appearance | Clear light yellow to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5650 to 1.5700 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...