2-Brom-6-methylpyridin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-Bromo-2-picoline, 2-Bromo-6-picoline

L20018.06 L20018.14
L20018.06EA 82.2 EUR
L20018.06 L20018.14
2-Brom-6-methylpyridin ≥98%
2-Brom-6-methylpyridin

Purity: 98% CAS No.: 5315-25-3 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 97°(206°F) Refractive Index: 1.5625 Boiling Point: 102-103°/20mm Beilstein Reference: 107322 MDL No.: MFCD00040743

Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
Siedepunkt: 102…103 °C (20 mmHg)
Dichte: 1.512
Flammpunkt: 97 °C (206 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040743
CAS-Nummer: 5315-25-3
EINECS: 226-173-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR