2-Brom-6-methylpyridin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 6-Bromo-2-picoline, 2-Bromo-6-picoline

020192-25G 020192-5G
FLUO020192-25GEA 60.9 EUR
FLUO020192-25G FLUO020192-5G
2-Brom-6-methylpyridin
2-Brom-6-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
MDL: MFCD00040743
CAS-Nummer: 5315-25-3
EINECS: 226-173-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR