2-Chlor-6-methylpyridin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Chlor-2-picolin
B20174.06
B20174.14
B20174.06EA
62.1
EUR
B20174.06
B20174.14
2-Chlor-6-methylpyridin ≥98%
2-Chlor-6-methylpyridin
Purity: 98%
CAS No.: 18368-63-3
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Flash Point: 73°(163°F)
Refractive Index: 1.5265
Boiling Point: 64-68°/10mm
Beilstein Reference: 107187
UN No.: 2810
MDL No.: MFCD00006245
Formel:
C₆H₆ClN Molecular Weight: 127,57 g/mol Siedepunkt: 64…68 °C (10 mmHg) Dichte: 1,167 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006245 CAS-Nummer: 18368-63-3 EINECS: 242-241-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...